Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Plug, 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms
SDP
Catalog No. CG42405
Encompass_Preferred
2mm 120° PTFE Plug Only, 15.2/30. A 1:5 taper PTFE stopcock with three sidearms sealed 120º apart. Plug has a bore to allow connection of any two arms at once.
Catalog No. CG42405 Supplier Chemglass Life Sciences Supplier No. CG42405
2mm 120° PTFE Plug Only, 15.2/30. A 1:5 taper PTFE stopcock with three sidearms sealed 120º apart. Plug has a bore to allow connection of any two arms at once.