Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG42403
Encompass_Preferred
Stopcock, Straight Bore, PTFE Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. CG42403 Supplier Chemglass Life Sciences Supplier No. CG42403
Stopcock, Straight Bore, PTFE Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.