Learn More
BURKLE INC StopCock straight-through,cap nut3/4"-cap nut3/4"
Catalog No. 5000025838
Encompass_Preferred
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.
Catalog No. 50-000-25838 Supplier BURKLE INC Supplier No. 05257000
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.