Learn More
BURKLE INC StopCock straight-through, 3/4" outer - 3/4" outer
Catalog No. 5000025945
Encompass_Preferred
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.
Catalog No. 50-000-25945 Supplier BURKLE INC Supplier No. 05253000
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.