Learn More
Ace Glass, Inc. Stopcock, 3-way, 1mm bore, 17/40 T bore plug, 8mm OD sidearms
SDP
Catalog No. 50976129
Encompass_Preferred
1mm bore 3-way stopcock with sidearms and a 17/40 T-bore solid glass plug. 8mm O.D.
Catalog No. 50-976-129 Supplier Ace Glass, Inc. Supplier No. 814502
1mm bore 3-way stopcock with sidearms and a 17/40 T-bore solid glass plug. 8mm O.D.