Learn More
Ace Glass, Inc. Stopcock, 3-way, 2mm bore, 12/30 T-bore plug, 8mm OD, sidearms
SDP
Catalog No. 50976128
Encompass_Preferred
2mm bore 3-way stopcock with sidearms and a 12/30 T-bore solid glass plug. 8mm O.D.
Catalog No. 50-976-128 Supplier Ace Glass, Inc. Supplier No. 814501
2mm bore 3-way stopcock with sidearms and a 12/30 T-bore solid glass plug. 8mm O.D.