Learn More
Ace Glass, Inc. Stopcock, 3-way, 3mm bore, 20/44 T-bore plug, 10mm OD, sidearms
SDP
Catalog No. 50979698
Encompass_Preferred
3mm bore 3-way stopcock with sidearms and a 20/44 T-bore solid glass plug. 10mm O.D.
Catalog No. 50-979-698 Supplier Ace Glass, Inc. Supplier No. 814506
3mm bore 3-way stopcock with sidearms and a 20/44 T-bore solid glass plug. 10mm O.D.