Learn More
Ace Glass, Inc. Stopcock, 3-way, 4mm bore, 17/40 T-bore plug, 10mm OD, sidearms
SDP
Catalog No. 50976131
Encompass_Preferred
4mm bore 3-way stopcock with sidearms and a 17/40 T-bore solid glass plug. 10mm O.D.
Catalog No. 50-976-131 Supplier Ace Glass, Inc. Supplier No. 814507
4mm bore 3-way stopcock with sidearms and a 17/40 T-bore solid glass plug. 10mm O.D.