Learn More
Chemglass Life Sciences Stopcock, T-Bore, Glass Plug, 4mm Bore, 20/44 Plug Size, 12mm OD Sidearms, #5 Clip Size
SDP
Catalog No. CG41004
Encompass_Preferred
Stopcock, T-Bore, Glass Plug, 4mm Bore, 20/44 Plug Size, 12mm OD Sidearms, #5 Clip Size. Stopcock has three sidearms sealed 90º apart. The solid glass plug has a "T" shaped bore so that any two or all three sidearms can be connected at one time. All sizes are supplied complete with a metal retaining device CG-423.
Catalog No. CG41004 Supplier Chemglass Life Sciences Supplier No. CG41004
Stopcock, T-Bore, Glass Plug, 4mm Bore, 20/44 Plug Size, 12mm OD Sidearms, #5 Clip Size. Stopcock has three sidearms sealed 90º apart. The solid glass plug has a "T" shaped bore so that any two or all three sidearms can be connected at one time. All sizes are supplied complete with a metal retaining device CG-423.