Learn More
AVANTOR MASTERFLEX (VWR LLC) PVC Stopcock w/ Flow Indication, 1/4-1/2"" ID x 1/4-1/2"" ID
Catalog No. NC9642437
Encompass
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120¿>F (49¿>C); maximum vacuum is 29.9'' Hg.
Catalog No. NC9642437 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0622556
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120¿>F (49¿>C); maximum vacuum is 29.9'' Hg.