Learn More
Accuform™ STOPOUT™ Universal Blockout Wall Switch Cover
Catalog No. 19211292 Shop All AccuformNMC Products
Catalog No. 19-211-292 Supplier Accuform™ Supplier No. KDD139
Add to Cart
Add to Cart
The STOPOUT™ Universal Blockout Wall Switch Cover attaches to existing switch plate screw(s) and fully encloses the entire switch
  • Attaches to existing switch plate screw(s) and fully encloses the entire switch
  • Works on both rocker/toggle and flat paddle switches
  • Unique design reduces error – applies only in direction of desired lockout position – on or off
  • Cover folds down to block screws and switch access - secure with a padlock (sold separately)
Specifications
Product Type Wall Switch Cover