Learn More
Electron Microscopy Sciences Stoppered Bottle Clear Borosilicate, 1000 ml, 110 x 215 mm
SDP
Catalog No. 5027452
Encompass_Preferred
New designed Ground Glass Stoppered Bottles feature a no-drip pour lid for clean pouring of contents.  The most preferred container for storage of prepared chemicals, and stock solutions.  Octagon-shaped ground glass stopper is included.  Borosilicate '400' glass offers a high degree of protection from leaching.
Volume: 1000 ml
Size: 110 x 215 mm
Cone: 29/32
Catalog No. 50-274-52 Supplier Electron Microscopy Sciences Supplier No. 64206C
New designed Ground Glass Stoppered Bottles feature a no-drip pour lid for clean pouring of contents.  The most preferred container for storage of prepared chemicals, and stock solutions.  Octagon-shaped ground glass stopper is included.  Borosilicate '400' glass offers a high degree of protection from leaching.
Volume: 1000 ml
Size: 110 x 215 mm
Cone: 29/32