Learn More
Electron Microscopy Sciences Stoppered Bottle Clear Borosilicate, 5000 ml, 175 x 320 mm
SDP
Catalog No. 5027454
Encompass_Preferred
New designed Ground Glass Stoppered Bottles feature a no-drip pour lid for clean pouring of contents.  The most preferred container for storage of prepared chemicals, and stock solutions.  Octagon-shaped ground glass stopper is included.  Borosilicate '400' glass offers a high degree of protection from leaching.
Volume: 5000 ml
Size: 175 x 320 mm
Cone: 45/40
Catalog No. 50-274-54 Supplier Electron Microscopy Sciences Supplier No. 64209C
New designed Ground Glass Stoppered Bottles feature a no-drip pour lid for clean pouring of contents.  The most preferred container for storage of prepared chemicals, and stock solutions.  Octagon-shaped ground glass stopper is included.  Borosilicate '400' glass offers a high degree of protection from leaching.
Volume: 5000 ml
Size: 175 x 320 mm
Cone: 45/40