Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Cap Style Stopper
GREENER_CHOICESDP

Catalog No. 31501891 Shop All Kemtech Products
Change view
Click to view available options
Size:
14/20
24/40
29/42
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-891 29/42
31-501-889 14/20
31-501-890 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-891 Supplier Synthware™ Supplier No. S392942
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For an apparatus with an inner opening.

Specifications

Stopper Number 29/42
Autoclavable Autoclavable
Diameter (Metric) Bottom 32 mm
Diameter (Metric) Outer 32 mm
Diameter (Metric) Top 32 mm
Product Type Solid Stopper
Shape Rounded
Sterility Non-sterile
Color Clear
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
Quantity 1 Ea.
Size 29/42
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.