Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarstedt Inc STOPPER F/TUBE 23.5MM/PK1000

Catalog No. NC9222507 Shop All Sarstedt Inc. Products
Encompass_Preferred

Stopper for Tube 23.5mm Neutral

Catalog No. NC9222507 Supplier Sarstedt Inc Supplier No. 65.790
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stopper for Tube 23.5mm Neutral

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.