Learn More
Chemglass Life Sciences Stopper, Hex Head, Hollow, 24/40
SDP
Catalog No. CG300601
Encompass_Preferred
Stopper, Hex Head, Hollow, 24/40. Hexagonal shaped head of stopper prevents it from rolling off of the bench top. Flat surface will also permit stopper to be inverted and placed on end, thereby preventing possible contamination of ground surface.
Catalog No. CG300601 Supplier Chemglass Life Sciences Supplier No. CG300601
Stopper, Hex Head, Hollow, 24/40. Hexagonal shaped head of stopper prevents it from rolling off of the bench top. Flat surface will also permit stopper to be inverted and placed on end, thereby preventing possible contamination of ground surface.