Learn More
Hach Company Stopper, Polyethylene, Hollow, Size 2, 6/pk
Supplier:  Hach Company 1448001
Encompass
Made of low-density polyethylene. Interchangeable with rubber stoppers. Safer and more economical than glass. Fits snugly. Excellent as sample cup for pH meters, flame photometers and automatic analyzers, or as a small beaker. 25-26 mm (1") high.
Catalog No. 50-199-5044