Learn More
Chemglass Life Sciences 24/40 PTFE Hollow Stopper, 1/4" NPT, Complete with Extraction Nut
SDP
Catalog No. 50143622
Encompass_Preferred
24/40 PTFE Hollow Stopper, 1/4" NPT, Complete with Extraction Nut. PTFE stopper is similar to CG-3002-L but with the addition of one female NPT port in the center for connection of compression fittings or pressure/vacuum gauges. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.
Catalog No. 50-143-622 Supplier Chemglass Life Sciences Supplier No. CG3002K24
24/40 PTFE Hollow Stopper, 1/4" NPT, Complete with Extraction Nut. PTFE stopper is similar to CG-3002-L but with the addition of one female NPT port in the center for connection of compression fittings or pressure/vacuum gauges. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.