Learn More
Ace Glass, Inc. Stopper, impinger, 12/5 socket inlet, 12/5 ball outlet, 24/40, open tube end nozzle
SDP
Catalog No. 50974993
Encompass_Preferred
24/40 midget impinger stopper with 12/5 socket inlet and 12/5 ball outlet. Features an open tube end nozzle.
Catalog No. 50-974-993 Supplier Ace Glass, Inc. Supplier No. 753136
24/40 midget impinger stopper with 12/5 socket inlet and 12/5 ball outlet. Features an open tube end nozzle.