Learn More
Chemglass Life Sciences Stopper, PTFE, Minum-Ware
SDP
Catalog No. 501532691
Encompass_Preferred
Stopper, PTFE, Minum-Ware. Solid PTFE stopper for use with all 14/10 microscale components.
Catalog No. 50-153-2691 Supplier Chemglass Life Sciences Supplier No. MW6001
Stopper, PTFE, Minum-Ware. Solid PTFE stopper for use with all 14/10 microscale components.