Learn More
Chemglass Life Sciences Stopper, #15 O-Ring Joint, Airfree, Schlenk
SDP
Catalog No. AF021001
Encompass_Preferred
Stopper, #15 O-Ring Joint, Airfree, Schlenk. For use with the AF-0060 or AF-0061 manifolds or any Airfree component with an o-ring as a closure. Supplied complete with one Viton O-Ring.
Catalog No. AF021001 Supplier Chemglass Life Sciences Supplier No. AF021001
Stopper, #15 O-Ring Joint, Airfree, Schlenk. For use with the AF-0060 or AF-0061 manifolds or any Airfree component with an o-ring as a closure. Supplied complete with one Viton O-Ring.