Learn More
Ace Glass, Inc. Stopper only, Smog bubbler, 24/40, 8mm OD tube with 1.5mm ID capillary orifice tapered tip, 12/5 outer inlet arm joint, 12/5 inner outlet arm joint
SDP
Catalog No. 50979317
Encompass_Preferred
Stopper only, Smog bubbler, 24/40, 8mm OD tube with 1.5mm ID capillary orifice tapered tip, 12/5 outer inlet arm joint, 12/5 inner outlet arm joint
Catalog No. 50-979-317 Supplier Ace Glass, Inc. Supplier No. 752914
Stopper only, Smog bubbler, 24/40, 8mm OD tube with 1.5mm ID capillary orifice tapered tip, 12/5 outer inlet arm joint, 12/5 inner outlet arm joint