Learn More
Hach Company STOPPER, PENNY HEAD
Supplier:  Hach Company 2655900
Encompass
Replacement ground glass stopper. For use with the Cold Vapor Mercury Test, 14/20 standard taper joint. 1/pk
Catalog No. 50-199-6447