Learn More
Wilmad Labglass STOPPER PENNYHEAD 29/42
Catalog No. 50807827
Encompass_Preferred
Stopper, Pennyhead, Hollow, Full Length [ST] Inner Joint. Joint Size: [ST] 29/42. Hollow Pennyhead Stopper with closed bottom.
Catalog No. 50-807-827 Supplier Wilmad Labglass Supplier No. LG10300114
Stopper, Pennyhead, Hollow, Full Length [ST] Inner Joint. Joint Size: [ST] 29/42. Hollow Pennyhead Stopper with closed bottom.