Learn More
Chemglass Life Sciences 14/20 Stopper, Solid
SDP
Catalog No. CG3002L14
Encompass_Preferred
14/20 Stopper, Solid. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.
Catalog No. CG3002L14 Supplier Chemglass Life Sciences Supplier No. CG3002L14
14/20 Stopper, Solid. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.