Learn More
Chemglass Life Sciences 19/22 Stopper, Solid
SDP
Catalog No. 50873409
Encompass_Preferred
19/22 Stopper, Solid. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.
Catalog No. 50-873-409 Supplier Chemglass Life Sciences Supplier No. CG3002L19
19/22 Stopper, Solid. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.