Learn More
Chemglass Life Sciences 45/50 Stopper, Hollow
SDP
Catalog No. CG3002L45
Encompass_Preferred
45/50 Stopper, Hollow. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.
Catalog No. CG3002L45 Supplier Chemglass Life Sciences Supplier No. CG3002L45
45/50 Stopper, Hollow. PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint. The 14/20 and 19/22 are solid, while the larger sizes are hollow.