Learn More
Thomas Scientific Stopper, Schoniger, 1000 mL
SDP
Catalog No. NC0837106
Encompass_Preferred
6513C60 stopper was traditionally offered by Thomas for 300/500 mL combustion flasks; glass tip extends 50 mm below ground joint 6513C65 stopper is used for 1,000 mL combustion flasks; glass tip extends 90 mm below ground joint All feature U-shaped platinum gauze with 3 mm opening for insertion of the sample wrapper. For replacement Platinum Carrier, see Thomas number 6514F55.
Catalog No. NC0837106 Supplier Thomas Scientific Supplier No. 6513C65
6513C60 stopper was traditionally offered by Thomas for 300/500 mL combustion flasks; glass tip extends 50 mm below ground joint 6513C65 stopper is used for 1,000 mL combustion flasks; glass tip extends 90 mm below ground joint All feature U-shaped platinum gauze with 3 mm opening for insertion of the sample wrapper. For replacement Platinum Carrier, see Thomas number 6514F55.