Learn More
Grainger Stopper Sz 0 9 mm BDia Black 0 PK181
Catalog No. 502766528
Encompass_Preferred
The EPDM Rubber Stopper from Grainger is a rubber stopper made from EPDM rubber. It has a diameter of 1-1/2" and a length of 1-1/2". The stopper has a solid top and no hole. It is rated for temperatures up to 250°F. The stopper provides an economical plug for closing off pipes and fittings. It creates an airtight and watertight seal. The EPDM rubber offers resistance to weather, ozone, aging, and chemicals. This stopper can be used in a variety of industrial applications.
Catalog No. 50-276-6528 Supplier Grainger Supplier No. 16UC45
The EPDM Rubber Stopper from Grainger is a rubber stopper made from EPDM rubber. It has a diameter of 1-1/2" and a length of 1-1/2". The stopper has a solid top and no hole. It is rated for temperatures up to 250°F. The stopper provides an economical plug for closing off pipes and fittings. It creates an airtight and watertight seal. The EPDM rubber offers resistance to weather, ozone, aging, and chemicals. This stopper can be used in a variety of industrial applications.