Learn More
Grainger Stopper Sz 1 14 mm BDia Black 0 PK65
Catalog No. 502765206
Encompass_Preferred
The product is a 16VA53 transformer by Hammond Manufacturing. It is a single phase transformer with a primary voltage of 120V and secondary voltage of 24V. The transformer has a capacity of 16VA and can output a maximum secondary current of 0.67A. It features an open frame design and measures approximately 2.4" x 2.2" x 2.4". The transformer has an operating temperature range of -20°C to 40°C and is RoHS compliant. It is designed for PCB mounting and low voltage control applications. The product is made in Canada and comes with a 5 year warranty from Hammond Manufacturing.
Catalog No. 50-276-5206 Supplier Grainger Supplier No. 16VA53
The product is a 16VA53 transformer by Hammond Manufacturing. It is a single phase transformer with a primary voltage of 120V and secondary voltage of 24V. The transformer has a capacity of 16VA and can output a maximum secondary current of 0.67A. It features an open frame design and measures approximately 2.4" x 2.2" x 2.4". The transformer has an operating temperature range of -20°C to 40°C and is RoHS compliant. It is designed for PCB mounting and low voltage control applications. The product is made in Canada and comes with a 5 year warranty from Hammond Manufacturing.