Learn More
Grainger Stopper Sz 11 48 mm BDia Black 0 PK7
Catalog No. 502767420
Encompass_Preferred
The Grainger 16VC08 is a 16-ounce aerosol can of CRC Brakleen non-chlorinated brake parts cleaner. It is a fast drying, non-flammable cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It contains no 1,1,1 trichloroethane, chlorofluorocarbons, or any ozone depleting substances. This brake parts cleaner features a 360-degree valve for convenient application at any angle.
Catalog No. 50-276-7420 Supplier Grainger Supplier No. 16VC08
The Grainger 16VC08 is a 16-ounce aerosol can of CRC Brakleen non-chlorinated brake parts cleaner. It is a fast drying, non-flammable cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It contains no 1,1,1 trichloroethane, chlorofluorocarbons, or any ozone depleting substances. This brake parts cleaner features a 360-degree valve for convenient application at any angle.