Learn More
Grainger Stopper Sz 11 48 mm BDia Black 0 PK7
Catalog No. 502766281
Encompass_Preferred
The Grainger 16VC09 is a 16-ounce aerosol can of CRC Brakleen non-chlorinated brake parts cleaner. It is a fast drying, non-flammable spray that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. The aerosol spray reaches hard to access areas.
Catalog No. 50-276-6281 Supplier Grainger Supplier No. 16VC09
The Grainger 16VC09 is a 16-ounce aerosol can of CRC Brakleen non-chlorinated brake parts cleaner. It is a fast drying, non-flammable spray that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. The aerosol spray reaches hard to access areas.