Learn More
Grainger Stopper Sz 11.5 50 mm BDia Black 0 PK6
Catalog No. 502765210
Encompass_Preferred
The Grainger 16VC11 is a single-phase, open drip-proof electric motor with a 1/3 horsepower rating. It operates on 115V AC power at 1725 RPM with a 48-56 frame size. This motor has a C-face endshield for easy mounting and features a cast iron frame and aluminum endshields. It can be used for various light-duty industrial applications.
Catalog No. 50-276-5210 Supplier Grainger Supplier No. 16VC11
The Grainger 16VC11 is a single-phase, open drip-proof electric motor with a 1/3 horsepower rating. It operates on 115V AC power at 1725 RPM with a 48-56 frame size. This motor has a C-face endshield for easy mounting and features a cast iron frame and aluminum endshields. It can be used for various light-duty industrial applications.