Learn More
Grainger Stopper Sz 11.5 50 mm BDia Black 0 PK6
Catalog No. 502765209
Encompass_Preferred
The 16VC10 is a 10-gallon, vertical, oil-lubricated air compressor from Ingersoll Rand. It has a 2-stage pump that delivers 16 CFM at 175 PSI with a maximum pressure of 175 PSI. The motor is 5 HP, 230V, 1 phase. It has a cast iron cylinder and an ASME coded air receiver tank. The compressor includes an aftercooler, belt guard, and automatic tank drain. This air compressor is designed for automotive service, body shops, fleet maintenance, machine shops, production lines, wood working, dry cleaners, car washes, general maintenance/repair shops, tire shops, and other light industrial applications. The 16VC10 provides a reliable supply of quality air for air tools and other pneumatic equipment.
Catalog No. 50-276-5209 Supplier Grainger Supplier No. 16VC10
The 16VC10 is a 10-gallon, vertical, oil-lubricated air compressor from Ingersoll Rand. It has a 2-stage pump that delivers 16 CFM at 175 PSI with a maximum pressure of 175 PSI. The motor is 5 HP, 230V, 1 phase. It has a cast iron cylinder and an ASME coded air receiver tank. The compressor includes an aftercooler, belt guard, and automatic tank drain. This air compressor is designed for automotive service, body shops, fleet maintenance, machine shops, production lines, wood working, dry cleaners, car washes, general maintenance/repair shops, tire shops, and other light industrial applications. The 16VC10 provides a reliable supply of quality air for air tools and other pneumatic equipment.