Learn More
Grainger Stopper Sz 14 75 mm BDia Black 0
Catalog No. 502764321
Encompass_Preferred
The EPDM Rubber Stopper from Grainger is a rubber stopper made from EPDM rubber. It has a diameter of 1-1/2" and a height of 1-1/2". The stopper has a solid top and no hole. It is rated for temperatures up to 250°F. The stopper provides a tight seal and is resistant to acids, caustics, and ozone. It is suitable for use in pipelines, chemical tanks, and other industrial applications requiring a reliable seal. The stopper meets ASTM D2000 standards. It is sold individually.
Catalog No. 50-276-4321 Supplier Grainger Supplier No. 16WC47
The EPDM Rubber Stopper from Grainger is a rubber stopper made from EPDM rubber. It has a diameter of 1-1/2" and a height of 1-1/2". The stopper has a solid top and no hole. It is rated for temperatures up to 250°F. The stopper provides a tight seal and is resistant to acids, caustics, and ozone. It is suitable for use in pipelines, chemical tanks, and other industrial applications requiring a reliable seal. The stopper meets ASTM D2000 standards. It is sold individually.