Learn More
Grainger Stopper Sz 14 75 mm BDia Black 0
Catalog No. 502764319
Encompass_Preferred
The 16WC45 is a 16-inch diameter, 45-degree elbow made of carbon steel. It has a schedule 40 wall thickness and is rated for 150 psi working pressure at 100 degrees Fahrenheit. The elbow has beveled ends and is made in the USA. It conforms to ASTM A234 standards for carbon steel pipe fittings. The elbow is typically used in steam, water, air and other low pressure applications. It provides a cost-effective way to make 45 degree turns in piping systems.
Catalog No. 50-276-4319 Supplier Grainger Supplier No. 16WC45
The 16WC45 is a 16-inch diameter, 45-degree elbow made of carbon steel. It has a schedule 40 wall thickness and is rated for 150 psi working pressure at 100 degrees Fahrenheit. The elbow has beveled ends and is made in the USA. It conforms to ASTM A234 standards for carbon steel pipe fittings. The elbow is typically used in steam, water, air and other low pressure applications. It provides a cost-effective way to make 45 degree turns in piping systems.