Learn More
Grainger Stopper Sz 5 23 mm BDia Black 0 PK25
Catalog No. 502768207
Encompass_Preferred
The Grainger Approved 5-1H Stopper is a rubber stopper with a size 5 and 23mm bottom diameter. It is made of black rubber and has a height of 1 inch. This stopper fits openings from 0.984 to 1.063 inches and is suitable for use as a closure for bottles, jars, tubes and other containers. It provides an air-tight seal and the one piece molded construction resists cracking and deterioration. The stopper meets FDA regulations for food contact.
Catalog No. 50-276-8207 Supplier Grainger Supplier No. 16ZD50
The Grainger Approved 5-1H Stopper is a rubber stopper with a size 5 and 23mm bottom diameter. It is made of black rubber and has a height of 1 inch. This stopper fits openings from 0.984 to 1.063 inches and is suitable for use as a closure for bottles, jars, tubes and other containers. It provides an air-tight seal and the one piece molded construction resists cracking and deterioration. The stopper meets FDA regulations for food contact.