Learn More
Grainger Stopper Sz 5 23 mm BDia Black 0 PK25
Catalog No. 502767840
Encompass_Preferred
The Grainger Approved Stopper Sz 5, 23 mm BDia, Black is a rubber stopper with an overall diameter of 23mm and a body diameter of 5mm. It is made of black rubber and has a solid top design. The stopper is used to plug openings in laboratory glassware and features a tapered design for a tight fit. It provides an airtight and watertight seal and is resistant to acids, bases, and most solvents. The stopper is rated to withstand temperatures up to 200°F. It meets ASTM D3574 standards and is an economical option for basic laboratory applications. The package contains 100 stoppers.
Catalog No. 50-276-7840 Supplier Grainger Supplier No. 16ZD49
The Grainger Approved Stopper Sz 5, 23 mm BDia, Black is a rubber stopper with an overall diameter of 23mm and a body diameter of 5mm. It is made of black rubber and has a solid top design. The stopper is used to plug openings in laboratory glassware and features a tapered design for a tight fit. It provides an airtight and watertight seal and is resistant to acids, bases, and most solvents. The stopper is rated to withstand temperatures up to 200°F. It meets ASTM D3574 standards and is an economical option for basic laboratory applications. The package contains 100 stoppers.