Learn More
Grainger Stopper Sz 8 33 mm BDia Black 0 PK12
Catalog No. 502767777
Encompass_Preferred
The Grainger 16ZD71 is a Westward brand 1/4-inch drive socket set. It includes 17 sockets ranging from 4mm to 14mm in both standard and deep configurations. The sockets are made from chrome vanadium steel and have a matte black phosphate finish. The set comes packaged in a molded case for storage and portability. This socket set is ideal for automotive and machinery applications that require metric sized fasteners. The lifetime warranty provides peace of mind that the sockets are a quality and durable product.
Catalog No. 50-276-7777 Supplier Grainger Supplier No. 16ZD71
The Grainger 16ZD71 is a Westward brand 1/4-inch drive socket set. It includes 17 sockets ranging from 4mm to 14mm in both standard and deep configurations. The sockets are made from chrome vanadium steel and have a matte black phosphate finish. The set comes packaged in a molded case for storage and portability. This socket set is ideal for automotive and machinery applications that require metric sized fasteners. The lifetime warranty provides peace of mind that the sockets are a quality and durable product.