Learn More
Grainger Stopper Sz 9 37 mm BDia Black 0 PK10
Catalog No. 502768210
Encompass_Preferred
The Grainger 16ZD77 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding. It has a large viewing area for increased visibility. The helmet provides shade levels from 5 to 13 adjustable via external knob. It has adjustable headgear for comfort and optimal fit. The helmet meets ANSI Z87.1 safety standards. The lithium battery powers up to 1500 hours of use. The Speedglas helmet provides reliable eye protection for welders.
Catalog No. 50-276-8210 Supplier Grainger Supplier No. 16ZD77
The Grainger 16ZD77 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding. It has a large viewing area for increased visibility. The helmet provides shade levels from 5 to 13 adjustable via external knob. It has adjustable headgear for comfort and optimal fit. The helmet meets ANSI Z87.1 safety standards. The lithium battery powers up to 1500 hours of use. The Speedglas helmet provides reliable eye protection for welders.