Learn More
Hach Company Stopper-Two Hole
Supplier:  Hach Company 2943241
Encompass
13 5 Two Hole stopper consists of a 13 5 SBR plug with 2 x 3/8 OD PP tube adapters for connection to a 10/20L vacuum rated PP carboy Ideal for use with Xenosep Solvent Recovery option 1pk
Catalog No. 50-002-07264