Learn More
Electron Microscopy Sciences Stopwatch
SDP
Catalog No. 50949349
Encompass_Preferred
Very accurate with a click stop control button. Large LCD display. Rugged ABS plastic case. It counts up to 60 minutes in 1/100th second resolution. It also has a split interval clock and calculator functions. Comes complete with a leatherette case.
Catalog No. 50-949-349 Supplier Electron Microscopy Sciences Supplier No. 72091
Very accurate with a click stop control button. Large LCD display. Rugged ABS plastic case. It counts up to 60 minutes in 1/100th second resolution. It also has a split interval clock and calculator functions. Comes complete with a leatherette case.