Learn More
Agilent Technologies Storage/Reaction Microplate, 24-Well, Polypropylene, Square Well, Pyramid Bottom, 25/Pk.
Catalog No. 50995855
Encompass
This 24-well storage and reaction microplate is constructed from high-quality polypropylene and designed for efficient laboratory fluid handling. Featuring square wells with pyramid bottoms, it is optimized for high-volume storage, assays, and collection applications. The microplate adheres to ANSI/SLAS standards to ensure compatibility with standard laboratory equipment.
  • Volume capacity of 11.32 mL per well with a 10.18 mL working volume
  • Manufactured in a DNase/RNase free environment under ISO 9001:2015 standards
  • Square well configuration with pyramid bottoms for optimal sample retrieval
  • Standard microplate dimensions of 127.76 mm x 85.47 mm
  • Plate height of 44.04 mm
  • Natural color finish
  • Supplied in a pack of 25
Catalog No. 50-995-855 Supplier Agilent Technologies Supplier No. 201272100
This 24-well storage and reaction microplate is constructed from high-quality polypropylene and designed for efficient laboratory fluid handling. Featuring square wells with pyramid bottoms, it is optimized for high-volume storage, assays, and collection applications. The microplate adheres to ANSI/SLAS standards to ensure compatibility with standard laboratory equipment.
  • Volume capacity of 11.32 mL per well with a 10.18 mL working volume
  • Manufactured in a DNase/RNase free environment under ISO 9001:2015 standards
  • Square well configuration with pyramid bottoms for optimal sample retrieval
  • Standard microplate dimensions of 127.76 mm x 85.47 mm
  • Plate height of 44.04 mm
  • Natural color finish
  • Supplied in a pack of 25