Learn More
Agilent Technologies Agilent storage/reaction microplate, 48-well (8 rows x 6 columns), polypropylene, 5 mL/rectangle well, pyramid bottoms, 44 mm height, 25/pk
Catalog No. 50995851
Encompass
Agilent storage/reaction microplate, 48-well (8 rows x 6 columns), polypropylene, 5 mL/rectangle well, pyramid bottoms, 44 mm height, 25/pk
Catalog No. 50-995-851 Supplier Agilent Technologies Supplier No. 201238100
Agilent storage/reaction microplate, 48-well (8 rows x 6 columns), polypropylene, 5 mL/rectangle well, pyramid bottoms, 44 mm height, 25/pk