Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies Storage/Reaction Microplate, 48-Well, Polypropylene, Natural, 15/Pk.

Catalog No. 50995850
Encompass

These 48-well storage, assay, and collection plates are manufactured from natural polypropylene. The plates feature a 48-well count with a rectangle well shape and pyramid bottom design. They meet all ANSI/SLAS microplate standards and are produced in a DNase/RNase-free environment with ISO 9001:2015 operations.

  • Well shape: 8R/6C rectangle
  • Bottom shape: Pyramid
  • Max well volume: 7.85 mL
  • Working volume: 7.06 mL
  • Dimensions: 127.76 mm x 85.47 mm
  • Plate height: 68.33 mm
  • Well depth: 63.50 mm
  • Material: Polypropylene
  • Quantity: 15 per pack
  • Suggested seal: 201156-100

Catalog No. 50-995-850 Supplier Agilent Technologies Supplier No. 201236100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These 48-well storage, assay, and collection plates are manufactured from natural polypropylene. The plates feature a 48-well count with a rectangle well shape and pyramid bottom design. They meet all ANSI/SLAS microplate standards and are produced in a DNase/RNase-free environment with ISO 9001:2015 operations.

  • Well shape: 8R/6C rectangle
  • Bottom shape: Pyramid
  • Max well volume: 7.85 mL
  • Working volume: 7.06 mL
  • Dimensions: 127.76 mm x 85.47 mm
  • Plate height: 68.33 mm
  • Well depth: 63.50 mm
  • Material: Polypropylene
  • Quantity: 15 per pack
  • Suggested seal: 201156-100

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.