Learn More
Agilent Technologies Storage/Reaction Microplate, 96-Well, Polypropylene, Square Well, 25/Pk.
Catalog No. 50995857
Encompass
These 96-well storage and reaction microplates are engineered from ultra-high purity, medical-grade polypropylene to provide low-binding characteristics, ensuring excellent recovery of DNA (greater than 94%) and protein (greater than 95%) samples. These chemical-resistant plates feature square wells with conical bottoms and are manufactured in a DNase/RNase-free, ISO 9001:2015 certified facility to support high-throughput applications in drug discovery and genomics.
  • Molded from chemical-resistant, medical-grade polymers for performance with aggressive solutions and solvents
  • Produced to ANSI/SLAS standards for compatibility with major automation platforms and benchtop devices
  • Tolerant of low temperatures down to -80 °C and autoclavable at 121 °C for 20 minutes
  • Sealable using heat seals, adhesive films, or cap mats
  • Ideal for serial titration of test molecules or cells
Catalog No. 50-995-857 Supplier Agilent Technologies Supplier No. 201276100
These 96-well storage and reaction microplates are engineered from ultra-high purity, medical-grade polypropylene to provide low-binding characteristics, ensuring excellent recovery of DNA (greater than 94%) and protein (greater than 95%) samples. These chemical-resistant plates feature square wells with conical bottoms and are manufactured in a DNase/RNase-free, ISO 9001:2015 certified facility to support high-throughput applications in drug discovery and genomics.
  • Molded from chemical-resistant, medical-grade polymers for performance with aggressive solutions and solvents
  • Produced to ANSI/SLAS standards for compatibility with major automation platforms and benchtop devices
  • Tolerant of low temperatures down to -80 °C and autoclavable at 121 °C for 20 minutes
  • Sealable using heat seals, adhesive films, or cap mats
  • Ideal for serial titration of test molecules or cells