Learn More
Agilent Technologies Storage and Reaction Microplate, 96-Well, Polypropylene, 2 mL, 25/Pk.
Catalog No. 50995853
Encompass
These 96-well storage and reaction microplates are designed for a variety of laboratory applications, including biological assays, filtrate collection, and low-temperature storage. They are molded from medical-grade, chemical-resistant polypropylene for reliable performance with aggressive solutions and solvents.
  • Manufactured to ANSI/SLAS standards for compatibility with major automation platforms and benchtop devices
  • Well volume of 2 mL with square wells and pyramid bottoms
  • Low temperature tolerant down to -80°C
  • Autoclavable at 121°C and 15 psi for 20 minutes
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
  • Sealable with heat seals, adhesive seals, or cap mats
  • Designed for serial titration of molecules or cells
  • Complementary fit with filter microplates for centrifugation stacking
Catalog No. 50-995-853 Supplier Agilent Technologies Supplier No. 201240100
These 96-well storage and reaction microplates are designed for a variety of laboratory applications, including biological assays, filtrate collection, and low-temperature storage. They are molded from medical-grade, chemical-resistant polypropylene for reliable performance with aggressive solutions and solvents.
  • Manufactured to ANSI/SLAS standards for compatibility with major automation platforms and benchtop devices
  • Well volume of 2 mL with square wells and pyramid bottoms
  • Low temperature tolerant down to -80°C
  • Autoclavable at 121°C and 15 psi for 20 minutes
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
  • Sealable with heat seals, adhesive seals, or cap mats
  • Designed for serial titration of molecules or cells
  • Complementary fit with filter microplates for centrifugation stacking