Learn More
CELLTREAT Scientific Products Storage Box, 15mL Centrifuge Tubes, 36 Place, Polypropylene, Case Quantity 4
SDP
Catalog No. 5000670060
Encompass_Preferred
Conveniently store 15 and 50mL Centrifuge Tubes in temperatures as low as -80°C
Durable polypropylene Storage Boxes are fully autoclavable
Integrated rack features printed numerical coordinates
Safely and easily remove boxes from freezer racks using textured pull tab
Catalog No. 50-006-70060 Supplier CELLTREAT Scientific Products Supplier No. 229946
Conveniently store 15 and 50mL Centrifuge Tubes in temperatures as low as -80°C
Durable polypropylene Storage Boxes are fully autoclavable
Integrated rack features printed numerical coordinates
Safely and easily remove boxes from freezer racks using textured pull tab