Learn More
USA Scientific Inc Storage Box 5mL flip-cap tubes
Catalog No. 502744846
Encompass
Storage Box 5x5, holds up to 25 5.0 mL flip-cap tubes, 2.5 inches H, polypropylene, 4/pack
Catalog No. 50-274-4846 Supplier USA Scientific Inc Supplier No. 40140532
Storage Box 5x5, holds up to 25 5.0 mL flip-cap tubes, 2.5 inches H, polypropylene, 4/pack