Learn More
Chemglass Life Sciences Storage Box, Micro Centrifuge Tube, 64-Place
SDP
Catalog No. 501215204
Encompass_Preferred
Storage Box, Micro Centrifuge Tube, 64-Place. This micro centrifuge tube storage box with a polyurethane foam insert is ideal for holding up to 64 tubes from 0.5 to 2.0mL. A transparent cover allows you to see the contents of the box and is keyed to the base in order to prevent misalignment. To improve your inventory control, you can write with a pen or marker on the cover surface, which is printed with a series of squares, numbered from 1 to 64. You can also save space by stacking these boxes in freezers, refrigerators and lab counters.
Catalog No. 50-121-5204 Supplier Chemglass Life Sciences Supplier No. CLS4300064
Storage Box, Micro Centrifuge Tube, 64-Place. This micro centrifuge tube storage box with a polyurethane foam insert is ideal for holding up to 64 tubes from 0.5 to 2.0mL. A transparent cover allows you to see the contents of the box and is keyed to the base in order to prevent misalignment. To improve your inventory control, you can write with a pen or marker on the cover surface, which is printed with a series of squares, numbered from 1 to 64. You can also save space by stacking these boxes in freezers, refrigerators and lab counters.